4-(2-Methoxyethyl)phenol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

B23548.09 B23548.18
B23548.09EA 43.7 EUR
B23548.09 B23548.18
4-(2-Methoxyethyl)phenol ≥98%
4-(2-Methoxyethyl)phenol

Purity: 98% CAS No.: 56718-71-9 Molecular Formula: C9H12O2 Formula Weight: 152.19 Melting Point: 43-45° MDL No.: MFCD00017537

Formel: C₉H₁₂O₂
Schmelzpunkt: 43…45 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017537
CAS-Nummer: 56718-71-9
EINECS: 260-354-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR