3-Sulfobenzoesäure Mononatriumsalz ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Sodium 3-sulphobenzoate

B23556.14 B23556.22 B23556.36
B23556.14EA 26.6 EUR
B23556.14 B23556.22 B23556.36
3-Sulfobenzoesäure Mononatriumsalz ≥97%
3-Sulfobenzoesäure Mononatriumsalz

Purity: 97% CAS No.: 17625-03-5 Molecular Formula: C7H5NaO5S Formula Weight: 224.17 MDL No.: MFCD00066378

Formel: C₇H₅NaO₅S
Lagertemperatur: Raumtemperatur
MDL: MFCD00066378
CAS-Nummer: 17625-03-5
EINECS: 241-602-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR