α-Brom-2-chlor-4-fluortoluol ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-Chlor-4-fluorbenzylbromid
B23565.06
B23565.14
B23565.06EA
17.2
EUR
B23565.06
B23565.14
α-Brom-2-chlor-4-fluortoluol ≥97%
α-Brom-2-chlor-4-fluortoluol
Purity: 97%
CAS No.: 45767-66-6
Molecular Formula: C7H5BrClF
Formula Weight: 223.48
Melting Point: 35-39°
Beilstein Reference: 3539265
UN No.: 3261
MDL No.: MFCD00236025
Formel:
C₇H₅BrClF Molecular Weight: 223.47 g/mol Schmelzpunkt: 35…39 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236025 CAS-Nummer: 45767-66-6 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...