HBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat) ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
HBTU, 1-(Bis(dimethylamino)methylene)-1H-benzo[d][1,2,3]triazole-1-ium 3-oxide hexafluorophosphate(V)
B23597.22
B23597.14
B23597.06
B23597.22EA
90.9
EUR
B23597.22
B23597.14
B23597.06
HBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat) ≥98%
HBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat)
Purity: 98%
CAS No.: 94790-37-1
Molecular Formula: C11H16F6N5OP
Formula Weight: 379.25
Storage Temperature: 2°C to 8°C
Melting Point: ca 200° dec.
Beilstein Reference: 7328329
MDL No.: MFCD00075445
Formel:
C₁₁H₁₆F₆N₅OP Molecular Weight: 379.2 g/mol Schmelzpunkt: ∼ 200 °C (Zers.) |
MDL:
MFCD00075445 CAS-Nummer: 94790-37-1 EINECS: 423-020-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...