Mangan(II)acetat, wasserfrei 98+%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B23708.0B
B23708.18
B23708.30
B23708.0BEA
422
EUR
B23708.0B
B23708.18
B23708.30
Mangan(II)acetat, wasserfrei 98+%
Mangan(II)acetat
Purity: 98+%
CAS No.: 638-38-0
Molecular Formula: C4H6MnO4
Formula Weight: 173.03
Merck Reference: 14,5724
MDL No.: MFCD00013039
Formel:
(H₃CCOO)₂Mn Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013039 CAS-Nummer: 638-38-0 EINECS: 211-334-3 Merck Index: 14,5724 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...