Mangan(II)acetat, wasserfrei 98+%

Lieferant: Thermo Fisher Scientific
Achtung

B23708.0B B23708.18 B23708.30
B23708.0BEA 422 EUR
B23708.0B B23708.18 B23708.30
Mangan(II)acetat, wasserfrei 98+%
Mangan(II)acetat

Purity: 98+% CAS No.: 638-38-0 Molecular Formula: C4H6MnO4 Formula Weight: 173.03 Merck Reference: 14,5724 MDL No.: MFCD00013039

Formel: (H₃CCOO)₂Mn
Lagertemperatur: Raumtemperatur
MDL: MFCD00013039
CAS-Nummer: 638-38-0
EINECS: 211-334-3
Merck Index: 14,5724

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR