Tetrakis(dimethylsiloxy)silan ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B23754.06
B23754.14
B23754.22
B23754.06EA
32
EUR
B23754.06
B23754.14
B23754.22
Tetrakis(dimethylsiloxy)silan ≥97%
Tetrakis(dimethylsiloxy)silan
Purity: 97%
CAS No.: 17082-47-2
Molecular Formula: C8H28O4Si5
Formula Weight: 328.73
Flash Point: 67°(153°F)
Refractive Index: 1.3880
Melting Point: -60°
Boiling Point: 188-190°
Beilstein Reference: 1793898
MDL No.: MFCD00053848
Formel:
C₈H₂₈O₄Si₅ Siedepunkt: 188…190 °C Schmelzpunkt: –60 °C Dichte: 0,884 Flammpunkt: 67 °C (153 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00053848 CAS-Nummer: 17082-47-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...