N-(tert-Butyldimethylsilyl)-N-methyltrifluoracetamid (MTBSTFA) ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

B23760.03 B23760.06 B23760.14
B23760.03EA 38.5 EUR
B23760.03 B23760.06 B23760.14
N-(tert-Butyldimethylsilyl)-N-methyltrifluoracetamid (MTBSTFA) ≥97%
N-(tert-Butyldimethylsilyl)-N-methyltrifluoracetamid (MTBSTFA)

Purity: 97% CAS No.: 77377-52-7 Molecular Formula: C9H18F3NOSi Formula Weight: 241.33 Flash Point: 52°(125°F) Refractive Index: 1.4020 Boiling Point: 168-170° Beilstein Reference: 3606546 UN No.: 1993 MDL No.: MFCD00009671

Formel: C₉H₁₈F₃NOSi
Siedepunkt: 168…170 °C
Dichte: 1,023
Flammpunkt: 52 °C (125 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009671
CAS-Nummer: 77377-52-7
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR