2-Amino-5-nitrobenzophenon 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: (2-Amino-5-nitrophenyl)(phenyl)methanone

B23773.22
B23773.22EA 134 EUR
B23773.22
2-Amino-5-nitrobenzophenon 98%
2-Amino-5-nitrobenzophenon

Purity: 98% CAS No.: 1775-95-7 Molecular Formula: C13H10N2O3 Formula Weight: 242.23 Melting Point: 166-168° MDL No.: MFCD00007364

Formel: C₁₃H₁₀N₂O₃
Molecular Weight: 242,23 g/mol
Schmelzpunkt: 166…168 °C
Dichte: 1,474 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007364
CAS-Nummer: 1775-95-7
EINECS: 217-207-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR