N-Acetyl-D-phenylalanin 99%

Lieferant: Thermo Fisher Scientific

Synonyme: Ac-Phe-OH

B23812.03 B23812.06 B23812.14
B23812.03EA 19.7 EUR
B23812.03 B23812.06 B23812.14
N-Acetyl-D-phenylalanin 99%
N-Acetyl-L-phenylalanine

Purity: 99% CAS No.: 2018-61-3 Molecular Formula: C11H13NO3 Formula Weight: 207.23 Melting Point: 166-170° Optical Rotation: +40° (c=1 in methanol) MDL No.: MFCD00063158

Formel: C₁₁H₁₃NO₃
Schmelzpunkt: 166…170 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00063158
CAS-Nummer: 2018-61-3
EINECS: 217-959-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR