4-Amino-2,3,5,6-tetrafluorpyridin 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2,3,5,6-Tetrafluoropyridin-4-amine
B23815.06
B23815.06EA
83
EUR
B23815.06
4-Amino-2,3,5,6-tetrafluorpyridin 98%
4-Amino-2,3,5,6-tetrafluoropyridine
Purity: 98%
CAS No.: 1682-20-8
Molecular Formula: C5H2F4N2
Formula Weight: 166.08
Melting Point: 84-87°
Beilstein Reference: 475870
Formel:
C₅H₂F₄N₂ Molecular Weight: 166,08 g/mol Schmelzpunkt: 84…87 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010855 CAS-Nummer: 1682-20-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...