Thymolphthalein

Lieferant: Thermo Fisher Scientific

Synonyme: 2-(1-Hydroxy-1,1-di(4-hydroxy-2-methyl-5-(methylethyl)phenyl)methyl)benzoesäurelacton, 3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalid

B23896.30 B23896.18 B23896.09
B23896.30EA 320 EUR
B23896.30 B23896.18 B23896.09
Thymolphthalein
Thymolphthalein

CAS No.: 125-20-2 Molecular Formula: C28H30O4 Formula Weight: 430.55 Melting Point: 251-253° Beilstein Reference: 359413 Merck Reference: 14,9401 MDL No.: MFCD00005909

Formel: C₂₈H₃₀O₄
Schmelzpunkt: 251…253 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005909
CAS-Nummer: 125-20-2
EINECS: 204-729-7
Merck Index: 14,9401

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR