1,3-Diisopropylbenzol 96%
Lieferant: Thermo Fisher Scientific
Achtung
|
B23904.18
B23904.30
B23904.18EA
31.1
EUR
B23904.18
B23904.30
1,3-Diisopropylbenzol 96%
1,3-Diisopropylbenzol
Purity: 96%
CAS No.: 99-62-7
Molecular Formula: C12H18
Formula Weight: 162.27
Flash Point: 76°(170°F)
Refractive Index: 1.4890
Melting Point: -63°
Boiling Point: 204°
Beilstein Reference: 1905828
MDL No.: MFCD00008889
Formel:
C₁₂H₁₈ Molecular Weight: 162.27 g/mol Siedepunkt: 204 °C Schmelzpunkt: –63 °C Dichte: 0.856 Flammpunkt: 76 °C (170 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008889 CAS-Nummer: 99-62-7 EINECS: 202-773-1 UN: 3082 ADR: 9 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...