meso-Hydrobenzoin 95%

Lieferant: Thermo Fisher Scientific

B23946.06 B23946.14
B23946.06EA 76 EUR
B23946.06 B23946.14
meso-Hydrobenzoin 95%
meso-Hydrobenzoin

Purity: 99% CAS No.: 579-43-1 Molecular Formula: C14H14O2 Formula Weight: 214.27 Melting Point: 134-136° Beilstein Reference: 2050814 MDL No.: MFCD00064253

Formel: C₁₄H₁₄O₂
Schmelzpunkt: 134…136 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064253
CAS-Nummer: 579-43-1
EINECS: 207-758-3
Merck Index: 14,4777

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR