3,6-Di-tert-butylfluoren ≥98%
Lieferant: Thermo Fisher Scientific
B23996.03
B23996.06
B23996.14
B23996.03EA
85.2
EUR
B23996.03
B23996.06
B23996.14
3,6-Di-tert-butylfluoren ≥98%
3,6-Di-tert-butylfluoren
Purity: 98%
CAS No.: 58775-07-8
Molecular Formula: C21H26
Formula Weight: 278.44
Melting Point: 79-82°
MDL No.: MFCD02683109
Formel:
C₂₁H₂₆ Schmelzpunkt: 79…82 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02683109 CAS-Nummer: 58775-07-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...