3,6-Di-tert-butylfluoren ≥98%

Lieferant: Thermo Fisher Scientific

B23996.03 B23996.06 B23996.14
B23996.03EA 85.2 EUR
B23996.03 B23996.06 B23996.14
3,6-Di-tert-butylfluoren ≥98%
3,6-Di-tert-butylfluoren

Purity: 98% CAS No.: 58775-07-8 Molecular Formula: C21H26 Formula Weight: 278.44 Melting Point: 79-82° MDL No.: MFCD02683109

Formel: C₂₁H₂₆
Schmelzpunkt: 79…82 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02683109
CAS-Nummer: 58775-07-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR