Sodium 2,4-pentanedionate hydrate 95%

Lieferant: Thermo Fisher Scientific
Achtung

B24009.0B B24009.18 B24009.30
B24009.0BEA 512 EUR
B24009.0B B24009.18 B24009.30
Sodium 2,4-pentanedionate hydrate 95%
Sodium 2,4-pentanedionate hydrate

Purity: 95% CAS No.: 86891-03-4 Molecular Formula: C5H7NaO2.xH2O Formula Weight: 122.11(anhy) Melting Point: 228-230° MDL No.: MFCD00078034

Formel: C₅H₇NaO₂·xH₂O
Schmelzpunkt: 228…230 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00078034
CAS-Nummer: 86891-03-4
EINECS: 239-448-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR