1,4-Bis(2-hydroxyethoxy)benzene 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
Hydroquinone Di(2-hydroxyethyl) Ether, 1,4-Di(2-hydroxyethoxy)benzene, Hydroquinone Bis(2-hydroxyethyl) Ether
B24037.0E
B24037.22
B24037.36
B24037.0EEA
285
EUR
B24037.0E
B24037.22
B24037.36
1,4-Bis(2-hydroxyethoxy)benzene 95%
1,4-Bis(2-hydroxyethoxy)benzene
Purity: 95%
CAS No.: 104-38-1
Molecular Formula: C10H14O4
Formula Weight: 198.22
Melting Point: 100-102°
MDL No.: MFCD00002861
Formel:
C₁₀H₁₄O₄ Molecular Weight: 198.22 g/mol Schmelzpunkt: 100…102 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002861 CAS-Nummer: 104-38-1 EINECS: 203-197-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...