5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazin-4,4''-disulfonsäure Mononatriumsalz Hydrat ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazin-4,4''-disulfonsäure Mononatriumsalz Hydrat

B24066.03 B24066.06 B24066.14
B24066.03EA 41.1 EUR
B24066.03 B24066.06 B24066.14
5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazin-4,4''-disulfonsäure Mononatriumsalz Hydrat ≥97%
Ferrozine mono-sodium salt hydrate

Purity: 97% CAS No.: 69898-45-9 Molecular Formula: C20H13N4NaO6S2.xH2O Formula Weight: 492.47(anhy) Beilstein Reference: 5222087 MDL No.: MFCD00150794

Formel: C₂₀H₁₃N₄NaO₆S₂·H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00150794
CAS-Nummer: 1266615-85-3
EINECS: 274-196-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR