Cobalt(II) citrate dihydrate 98%

Lieferant: Thermo Fisher Scientific
Gefahr

B24088.22 B24088.14
B24088.22EA 102 EUR
B24088.22 B24088.14
Cobalt(II) citrate dihydrate 98%
Cobalt(II) citrate dihydrate

Purity: 98% CAS No.: 6424-15-3 Molecular Formula: C12H10Co3O14.2H2O Formula Weight: 591.02 (554.99anhy) Melting Point: ca 150° dec. UN No.: UN3077 MDL No.: MFCD02684007

Formel: C₁₂H₁₀Co₃O₁₄·2H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD02684007
CAS-Nummer: 6424-15-3
EINECS: 212-751-3
ADR: 9,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR