4-Fluor-2-methylphenylboronsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Fluoro-2-methylbenzeneboronic acid, 2-Methyl-4-fluorophenylboronic acid

B24117.06 B24117.03 B24117.14
B24117.06EA 168 EUR
B24117.06 B24117.03 B24117.14
4-Fluor-2-methylphenylboronsäure ≥98%
4-Fluor-2-methylphenylboronsäure

Purity: 98% CAS No.: 139911-29-8 Molecular Formula: C7H8BFO2 Formula Weight: 153.95 Melting Point: 199-201° MDL No.: MFCD02093072

Formel: C₇H₈BFO₂
Molecular Weight: 153.95 g/mol
Schmelzpunkt: 199…201 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02093072
CAS-Nummer: 139911-29-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR