(Methylthio)aceton ≥98%
Lieferant: Thermo Fisher Scientific
Purity: 98+%
CAS No.: 14109-72-9
Molecular Formula: C4H8OS
Formula Weight: 104.17
Flash Point: 43°(109°F)
Refractive Index: 1.4740
Odour: Stench
Boiling Point: 69-70°/30mm
Beilstein Reference: 1737738
UN No.: 1224
MDL No.: MFCD00015325
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...