Hexadecacarbonylhexarhodium ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

B24185.02 B24185.77
B24185.02EA 794 EUR
B24185.02 B24185.77
Hexadecacarbonylhexarhodium ≥98%
Hexadecacarbonylhexarhodium

Purity: 98% CAS No.: 28407-51-4 Molecular Formula: Rh6(CO)16 Formula Weight: 1065.60 Melting Point: 300° UN No.: 3466 MDL No.: MFCD00011206

Formel: C₁₆O₁₆Rh₆
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011206
CAS-Nummer: 28407-51-4
EINECS: 249-009-3
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR