Phenolphthalein disulphate tripotassium salt hydrate 98%
Lieferant: Thermo Scientific
Synonyme:
Phenolphthalein disulphate potassium salt hydrate
B24198.03
B24198.03EA
65.3
EUR
B24198.03
Phenolphthalein disulphate tripotassium salt hydrate 98%
Phenolphthalein disulphate tripotassium salt hydrate
Purity: 99%
CAS No.: 62625-16-5
Molecular Formula: C20H13K3O11S2.xH2O
Formula Weight: 610.75(anhy)
MDL No.: MFCD00014730
Formel:
C₂₀H₁₄O₁₀S₂ Molecular Weight: 610,75 g/mol (anhy) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00070538 CAS-Nummer: 62625-16-5 EINECS: 263-648-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...