3,4-Dimethoxyphenylboronsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B24240.03
B24240.06
B24240.14
B24240.03EA
48.1
EUR
B24240.03
B24240.06
B24240.14
3,4-Dimethoxyphenylboronsäure ≥98%
3,4-Dimethoxyphenylboronsäure
Purity: 98%
CAS No.: 122775-35-3
Molecular Formula: C8H11BO4
Formula Weight: 181.98
Melting Point: ca 240°
Beilstein Reference: 5334877
Formel:
C₈H₁₁BO₄ Schmelzpunkt: ∼ 240 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01074574 CAS-Nummer: 122775-35-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...