(3,4,5-Trimethoxyphenyl)acetonitril ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 3,4,5-Trimethoxyphenylacetonitrile

B24247.06 B24247.14
B24247.06EA 43.5 EUR
B24247.06 B24247.14
(3,4,5-Trimethoxyphenyl)acetonitril ≥97%
(3,4,5-Trimethoxyphenyl)acetonitril

Purity: 97% CAS No.: 13338-63-1 Molecular Formula: C11H13NO3 Formula Weight: 207.23 Melting Point: 75-79° Beilstein Reference: 2214548 UN No.: 3439 MDL No.: MFCD00001912

Formel: C₁₁H₁₃NO₃
Molecular Weight: 207.23 g/mol
Schmelzpunkt: 75…79 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001912
CAS-Nummer: 13338-63-1
EINECS: 236-388-5
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR