2-Chlor-5-nitrobenzonitril ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

B24287.06 B24287.14 B24287.22
B24287.06EA 26.7 EUR
B24287.06 B24287.14 B24287.22
2-Chlor-5-nitrobenzonitril ≥99%
2-Chlor-5-nitrobenzonitril

Purity: 99% CAS No.: 16588-02-6 Molecular Formula: C7H3ClN2O2 Formula Weight: 182.57 Melting Point: 104-107° Beilstein Reference: 1818642

Formel: C₇H₃ClN₂O₂
Siedepunkt: 119…122 °C (0,6 mmHg)
Schmelzpunkt: 104…107 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007292
CAS-Nummer: 16588-02-6
EINECS: 240-643-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR