Cobalt(II) gluconate hydrate

Lieferant: Thermo Fisher Scientific
Gefahr

B24293.14 B24293.22
B24293.14EA 47 EUR
B24293.14 B24293.22
Cobalt(II) gluconate hydrate
Cobalt(II) gluconate hydrate

CAS No.: 71957-08-9 Molecular Formula: C12H22CoO14.xH2O Formula Weight: 449.23(anhy) Melting Point: 300° MDL No.: MFCD02684006

Formel: C₁₂H₂₂CoO₁₄·xH₂O
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02684006
CAS-Nummer: 1809162-99-9
EINECS: 276-206-1
UN: 3288
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR