ω-Brom-4-phenylacetophenon ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Biphenyl-4-ylbrommethylketon, α-Brom-4-phenylacetophenon, Brommethyl-p-phenylphenylketon, 4-Phenylphenacylbromid, 2-Brom-4'-phenylacetophenon

B24298.06 B24298.14 B24298.22
B24298.06EA 26 EUR
B24298.06 B24298.14 B24298.22
ω-Brom-4-phenylacetophenon ≥97%
ω-Brom-4-phenylacetophenon

Purity: 97% CAS No.: 135-73-9 Molecular Formula: C14H11BrO Formula Weight: 275.15 Melting Point: 123-127° Beilstein Reference: 513838 UN No.: 3261 MDL No.: MFCD00000202

Formel: C₁₄H₁₁BrO
Molecular Weight: 275.14 g/mol
Siedepunkt: 354
Schmelzpunkt: 123…127 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00000202
CAS-Nummer: 135-73-9
EINECS: 205-217-6
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR