Kongorot Indikator
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Direktrot 28, Direct Red 28, Dinatrium-3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalin-1-sulfonat)
B24310.14
B24310.22
B24310.36
B24310.14EA
47.7
EUR
B24310.14
B24310.22
B24310.36
Kongorot Indikator
Kongorot
CAS No.: 573-58-0
Molecular Formula: C32H22N6Na2O6S2
Formula Weight: 696.67
Odour: Odourless
Melting Point:
Formel:
C₃₂H₂₂N₆Na₂O₆S₂ Schmelzpunkt: >360 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004028 CAS-Nummer: 573-58-0 EINECS: 209-358-4 Merck Index: 14,2498 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...