Kongorot Indikator

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Direktrot 28, Direct Red 28, Dinatrium-3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalin-1-sulfonat)

B24310.14 B24310.22 B24310.36
B24310.14EA 47.7 EUR
B24310.14 B24310.22 B24310.36
Kongorot Indikator
Kongorot

CAS No.: 573-58-0 Molecular Formula: C32H22N6Na2O6S2 Formula Weight: 696.67 Odour: Odourless Melting Point: 360° Beilstein Reference: 3894858 Merck Reference: 14,2498 MDL No.: MFCD00004028

Formel: C₃₂H₂₂N₆Na₂O₆S₂
Schmelzpunkt: >360 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004028
CAS-Nummer: 573-58-0
EINECS: 209-358-4
Merck Index: 14,2498

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR