3-Chlorpyridin ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
B24316.09
B24316.18
B24316.09EA
30.7
EUR
B24316.09
B24316.18
3-Chlorpyridin ≥99%
3-Chloropyridine
Purity: 99%
CAS No.: 626-60-8
Molecular Formula: C5H4ClN
Formula Weight: 113.55
Flash Point: 53°(127°F)
Refractive Index: 1.5330
Boiling Point: 147-149°
Beilstein Reference: 105744
UN No.: 1993
MDL No.: MFCD00006375
Formel:
C₅H₄ClN Siedepunkt: 147…149 °C Dichte: 1,205 Flammpunkt: 53 °C (127 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006375 CAS-Nummer: 626-60-8 EINECS: 210-955-7 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...