2,3,5,6-Tetrafluor-4-(trifluormethyl)anilin ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Aminotetrafluorobenzotrifluoride
B24326.03
B24326.06
B24326.03EA
51.4
EUR
B24326.03
B24326.06
2,3,5,6-Tetrafluor-4-(trifluormethyl)anilin ≥98%
2,3,5,6-Tetrafluor-4-(trifluormethyl)anilin
Purity: 98%
CAS No.: 651-83-2
Molecular Formula: C7H2F7N
Formula Weight: 233.09
Refractive Index: 1.4300
Boiling Point: 185-187°
UN No.: 2810
MDL No.: MFCD00091518
Formel:
C₇H₂F₇N Molecular Weight: 233.09 g/mol Siedepunkt: 185…187 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00091518 CAS-Nummer: 651-83-2 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...