m-Kresolpurpur

Lieferant: Thermo Fisher Scientific

B24338.06 B24338.14
B24338.06EA 44.2 EUR
B24338.06 B24338.14
m-Kresolpurpur
m-Kresolpurpur

CAS No.: 2303-01-7 Molecular Formula: C21H18O5S Formula Weight: 382.44 Beilstein Reference: 350314 Merck Reference: 14,2577 MDL No.: MFCD00005871

Formel: C₂₁H₁₈O₅S
Lagertemperatur: Raumtemperatur
MDL: MFCD00005871
CAS-Nummer: 2303-01-7
EINECS: 218-960-6
Merck Index: 14,2577

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR