2-(Trifluormethyl)phenylboronsäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Dihydroxy(2-(trifluormethyl)phenyl)boran, 2-(Trifluormethyl)benzolboronsäure, (2-(Trifluormethyl)phenyl)metaborsäure
B24343.03
B24343.06
B24343.14
B24343.03EA
38.2
EUR
B24343.03
B24343.06
B24343.14
2-(Trifluormethyl)phenylboronsäure ≥97%
2-(Trifluormethyl)phenylboronsäure
Purity: 97%
CAS No.: 1423-27-4
Molecular Formula: C7H6BF3O2
Formula Weight: 189.93
Melting Point: 108-110°
Beilstein Reference: 2974465
Formel:
C₇H₆BF₃O₂ Schmelzpunkt: 108…110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236059 CAS-Nummer: 1423-27-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...