2-Chlor-4,6-dinitroanilin ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
6-Chlor-2,4-dinitroanilin
B24379.22
B24379.22EA
29
EUR
B24379.22
2-Chlor-4,6-dinitroanilin ≥97%
2-Chlor-4,6-dinitroanilin
Purity: 97%
CAS No.: 3531-19-9
Molecular Formula: C6H4ClN3O4
Formula Weight: 217.57
Melting Point: 153-155°
UN No.: 2811
MDL No.: MFCD00007147
Formel:
C₆H₄ClN₃O₄ Molecular Weight: 217,57 g/mol Schmelzpunkt: 153…155 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007147 CAS-Nummer: 3531-19-9 EINECS: 222-564-9 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...