5-Aminoindol 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
5-Indolamin, Indol-5-ylamin, 5-Amino-1H-indol
B24391.03
B24391.03EA
54.9
EUR
B24391.03
5-Aminoindol 97%
5-Aminoindol
Purity: 97%
CAS No.: 5192-03-0
Molecular Formula: C8H8N2
Formula Weight: 132.17
Storage Temperature: 2°C to 8°C
Melting Point: 130-134°
Beilstein Reference: 112348
MDL No.: MFCD00005679
Formel:
C₈H₈N₂ Molecular Weight: 132,16 g/mol Siedepunkt: 190 °C (6 mmHg) Schmelzpunkt: 130…134 °C |
MDL:
MFCD00005679 CAS-Nummer: 5192-03-0 EINECS: 225-977-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...