Ammonium cobalt(II) phosphate monohydrate 98%

Lieferant: Thermo Scientific
Gefahr

B24393.18
B24393.18EA 37.2 EUR
B24393.18
Ammonium cobalt(II) phosphate monohydrate 98%
Ammonium cobalt(II) phosphate monohydrate

Purity: 98% CAS No.: 16827-96-6 Molecular Formula: H4CoNO4P.H2O Formula Weight: 189.96 (171.94anhy) MDL No.: MFCD02683114

Formel: H₄CoNO₄P·H₂O
Molecular Weight: 189,96 g/mol (171,94 anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD02683114
CAS-Nummer: 16827-96-6
EINECS: 238-633-1
Merck Index: 14,513

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR