4-Brom-2,5-difluoranilin ≥98%

Lieferant: Thermo Scientific
Gefahr

B24457.09
B24457.09EA 78 EUR
B24457.09
4-Brom-2,5-difluoranilin ≥98%
4-Brom-2,5-difluoranilin

Purity: 98% CAS No.: 112279-60-4 Molecular Formula: C6H4BrF2N Formula Weight: 208.01 Melting Point: 74-75° Beilstein Reference: 4741102

Formel: C₆H₄BrF₂N
Molecular Weight: 208,01 g/mol
Schmelzpunkt: 74…75 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01861125
CAS-Nummer: 112279-60-4
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR