1,2-Propylendiamin 99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99%
CAS No.: 78-90-0
Molecular Formula: C3H10N2
Formula Weight: 74.13
Flash Point: 34°(93°F)
Refractive Index: 1.4460
Odour: Characteristic
Melting Point: ca -37°
Boiling Point: 119-120°
Beilstein Reference: 605274
Merck Reference: 14,7852
UN No.: 2258
MDL No.: MFCD00008089
Formel:
C₃H₁₀N₂ Siedepunkt: 119…120 °C Schmelzpunkt: ∼–37 °C Dichte: 0,869 Flammpunkt: 34 °C (93 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008089 CAS-Nummer: 78-90-0 EINECS: 201-155-9 UN: 2258 ADR: 8,II Merck Index: 14,7852 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...