5-Chlor-2-benzoxazolinon ≥99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 5-Chloro-2(3H)-benzoxazolone, 5-Chlorobenzo[d]oxazol-2(3H)-one, Chlorzoxazone, Chloroxazone

B24507.18
B24507.18EA 43.5 EUR
B24507.18
5-Chlor-2-benzoxazolinon ≥99%
5-Chlor-2-benzoxazolinon

Purity: 99% CAS No.: 95-25-0 Molecular Formula: C7H4ClNO2 Formula Weight: 169.57 Melting Point: 189-192° Merck Reference: 14,2194 MDL No.: MFCD00005717

Formel: C₇H₄ClNO₂
Molecular Weight: 169,57 g/mol
Schmelzpunkt: 189…192 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005717
CAS-Nummer: 95-25-0
EINECS: 202-403-9
Merck Index: 14,2194

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR