Iron(II) gluconate hydrate 94%

Lieferant: Thermo Fisher Scientific

B24561.22 B24561.36
B24561.22EA 22 EUR
B24561.22 B24561.36
Iron(II) gluconate hydrate 94%
Iron(II) gluconate hydrate

Purity: 94% CAS No.: 22830-45-1 Molecular Formula: C12H22FeO14.2H2O Formula Weight: 482.19 (446.19anhy) Melting Point: ca 140° MDL No.: MFCD00150872

Formel: C₁₂H₂₄FeO₁₅
Schmelzpunkt: ∼ 140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00150872
CAS-Nummer: 22830-45-1
EINECS: 206-076-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR