Suberoyl chloride 97%

Lieferant: Thermo Fisher Scientific
Gefahr

B24580.06 B24580.14
B24580.06EA 87.9 EUR
B24580.06 B24580.14
Suberoyl chloride 97%
Suberoyl chloride

Purity: 97% CAS No.: 10027-07-3 Molecular Formula: C8H12Cl2O2 Formula Weight: 211.09 Flash Point: 110°(230°F) Refractive Index: 1.4680 Boiling Point: 162-163°/15mm UN No.: UN3265 MDL No.: MFCD00000764

Formel: C₈H₁₂Cl₂O₂
Siedepunkt: 162…163 °C (15 mmHg)
Dichte: 1,172
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000764
CAS-Nummer: 10027-07-3
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR