2-Amino-4-hydroxy-6-methylpyrimidin 98%

Lieferant: Thermo Fisher Scientific

Synonyme: 2-Amino-6-methylpyrimidin-4(1H)-one, 2-Amino-6-methyl-4-pyrimidinol

B24604.18 B24604.30
B24604.18EA 38.3 EUR
B24604.18 B24604.30
2-Amino-4-hydroxy-6-methylpyrimidin 98%
2-Amino-4-hydroxy-6-methylpyrimidine

Purity: 98% CAS No.: 3977-29-5 Molecular Formula: C5H7N3O Formula Weight: 125.13 Melting Point: ca 305° dec. Beilstein Reference: 3531

Formel: C₅H₇N₃O
Molecular Weight: 125.13 g/mol
Schmelzpunkt: ∼ 305 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006095
CAS-Nummer: 3977-29-5
EINECS: 223-612-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR