4,4'-Dichlorbutyrophenon ≥97%

Lieferant: Thermo Fisher Scientific

B24608.14 B24608.18 B24608.22
B24608.14EA 50.2 EUR
B24608.14 B24608.18 B24608.22
4,4'-Dichlorbutyrophenon ≥97%
4,4'-Dichlorbutyrophenon

Purity: 97% CAS No.: 40877-09-6 Molecular Formula: C10H10Cl2O Formula Weight: 217.09 Refractive Index: 1.5600 Melting Point: 28-32° Beilstein Reference: 879782 MDL No.: MFCD00018993

Formel: C₁₀H₁₀Cl₂O
Siedepunkt: 135…140 °C (0,9 mmHg)
Schmelzpunkt: 28…32 °C
Dichte: 1,2575 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00018993
CAS-Nummer: 40877-09-6
EINECS: 255-123-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR