1,8-Octandithiol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

B24693.03 B24693.06 B24693.14
B24693.03EA 33.6 EUR
B24693.03 B24693.06 B24693.14
1,8-Octandithiol ≥98%
1,8-Octandithiol

Purity: 98% CAS No.: 1191-62-4 Molecular Formula: HS(CH2)8SH Formula Weight: 178.36 Flash Point: 113°(235°F) Refractive Index: 1.5050 Odour: Unpleasant Boiling Point: 269-270° Beilstein Reference: 1735431 MDL No.: MFCD00003574

Formel: C₈H₁₈S₂
Siedepunkt: 269…270 °C
Dichte: 0,960
Flammpunkt: 113 °C (235 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003574
CAS-Nummer: 1191-62-4
EINECS: 214-738-8
UN: 3334
ADR: 9,

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR