2,7-Dichlorfluoren ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2,7-Dichloro-9H-fluorene

B24717.06
B24717.06EA 112 EUR
B24717.06
2,7-Dichlorfluoren ≥97%
2,7-Dichlorfluoren

Purity: 97% CAS No.: 7012-16-0 Molecular Formula: C13H10Cl2 Formula Weight: 235.12 Melting Point: 300° dec. MDL No.: MFCD00032840

Formel: C₁₃H₈Cl₂
Molecular Weight: 235,11 g/mol
Schmelzpunkt: >300 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00032840
CAS-Nummer: 7012-16-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR