4-(Dimethylamino)zimtaldehyd ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

B24741.06 B24741.14
B24741.06EA 75 EUR
B24741.06 B24741.14
4-(Dimethylamino)zimtaldehyd ≥98%
4-(Dimethylamino)zimtaldehyd

Purity: 98% CAS No.: 6203-18-5 Molecular Formula: C11H13NO Formula Weight: 175.23 Melting Point: 137-144° Beilstein Reference: 972369

Formel: C₁₁H₁₃NO
Schmelzpunkt: 137…144 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007002
CAS-Nummer: 6203-18-5
EINECS: 228-267-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR