2-Ethoxyphenol ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Guaethol, 1-Hydroxy-2-ethoxybenzol, Brenzcatechinmonoethylether, o-Ethoxyphenol
B24755.36
B24755.36EA
98.8
EUR
B24755.36
2-Ethoxyphenol ≥98%
2-Ethoxyphenol
Purity: 98%
CAS No.: 94-71-3
Molecular Formula: C8H10O2
Formula Weight: 138.17
Flash Point: 86°(187°F)
Odour: Phenol-like
Boiling Point: 214-216°
MDL No.: MFCD00002187
Formel:
C₈H₁₀O₂ Molecular Weight: 138,17 g/mol Siedepunkt: 214…216 °C Schmelzpunkt: 24…27 °C Dichte: 1,10 Flammpunkt: 86 °C (187 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002187 CAS-Nummer: 94-71-3 EINECS: 202-358-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...