4-Benzyloxy-2-hydroxypyridin ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

B24774.06 B24774.14
B24774.06EA 103 EUR
B24774.06 B24774.14
4-Benzyloxy-2-hydroxypyridin ≥97%
4-Benzyloxy-2-hydroxypyridin

Purity: 97% CAS No.: 53937-02-3 Molecular Formula: C12H11NO2 Formula Weight: 201.22 Melting Point: 194-198° MDL No.: MFCD04039771

Formel: C₁₂H₁₁NO₂
Schmelzpunkt: 194…198 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00191783
CAS-Nummer: 53937-02-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR